Order Entry
Ireland
ContactUsLinkComponent
Quick Release Clamps for Flat Flange Vessels, Made of stainless steel, readjustable clamp with three retaining segments. 1 * 1 items
Catalog # BOHLB277-03
Supplier:  Bohlender
undefined
Quick Release Clamps for Flat Flange Vessels, Made of stainless steel, readjustable clamp with three retaining segments. 1 * 1 items
Catalog # BOHLB277-03
Supplier:  Bohlender
Frequently Bought Together