Order Entry
Ireland
ContactUsLinkComponent
High Select HSA/Immunoglobulin Depletion Resin decreases the albumin and antibody components that are abundant in human plasma samples, helping prepare those samples for mass spectrometry or 1D or 2D electrophoresis. The resin is provided here in a c 1 * 6 items
Catalog # PIERA36365
High Select HSA/Immunoglobulin Depletion Resin decreases the albumin and antibody components that are abundant in human plasma samples, helping prepare those samples for mass spectrometry or 1D or 2D electrophoresis. The resin is provided here in a c 1 * 6 items
Catalog # PIERA36365
Frequently Bought Together