Graduated cylinder, tall form, class A, PMP, 1000ml:10ml, DE-M marking, with batch certificate 1 * 1 items
Catalog # BRNDDAKKS35162
Supplier: Brand
Graduated cylinder, tall form, class A, PMP, 1000ml:10ml, DE-M marking, with batch certificate 1 * 1 items
Catalog # BRNDDAKKS35162
Supplier: Brand
Frequently Bought Together
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF
VWR® Reagent Reservoirs
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: EPPENDORF
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
VWR®, Forceps
Supplier: VWR Collection
Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Pipette Tips, Universal
Supplier: VWR Collection