EC HPLC column EC 100/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 100 mm, ID: 2 mm pack of 1 1 * 1 items
Catalog # MANA763434.20
Supplier: MACHEREY-NAGEL
EC HPLC column EC 100/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 100 mm, ID: 2 mm pack of 1 1 * 1 items
Catalog # MANA763434.20
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection