Pierce Anti-DYKDDDDK Magnetic Agarose provides a fast, convenient method for purification and immunoprecipitation (IP) of DYKDDDDK-tagged proteins from in vitro protein expression systems, bacteria, yeast, and mammalian cells. 1 * 1 mL
Catalog # PIERA36797
Supplier: Thermo Fisher Scientific
Frequently Bought Together