Order Entry
Ireland
ContactUsLinkComponent
Cone made from metal wire cloth Stainless steel wire 105 mm height and 100 mm diam w: 0.250 mm d: 0.160 mm 1 * 1 items
Catalog # HAVE205324352
Supplier:  NEXOPART
Cone made from metal wire cloth Stainless steel wire 105 mm height and 100 mm diam w: 0.250 mm d: 0.160 mm 1 * 1 items
Catalog # HAVE205324352
Supplier:  NEXOPART
Frequently Bought Together