Order Entry
Ireland
ContactUsLinkComponent
3/8' I.D. x 5/8' O.D. Molded Tee Liner Mainline: 1/2' TC Molded Flanges Outlet: 1/2' TC Molded Flange Material: Platinum Cured Silicone As Per Drawing # ART001285 1 * 1 items
Catalog # IBSAART001285
Supplier:  IBS Stock Items
3/8' I.D. x 5/8' O.D. Molded Tee Liner Mainline: 1/2' TC Molded Flanges Outlet: 1/2' TC Molded Flange Material: Platinum Cured Silicone As Per Drawing # ART001285 1 * 1 items
Catalog # IBSAART001285
Supplier:  IBS Stock Items
Frequently Bought Together