Order Entry
Ireland
ContactUsLinkComponent
Trichloroacetic acid (TCA) is used as a reagent for the precipitation of proteins and nucleic acids. Has been used to determine prot ein concentration by quantitative precipitation with the Lowry method. 1 * 1 kg
Catalog # GENORC-814
Supplier:  G-Biosciences
Trichloroacetic acid (TCA) is used as a reagent for the precipitation of proteins and nucleic acids. Has been used to determine prot ein concentration by quantitative precipitation with the Lowry method. 1 * 1 kg
Catalog # GENORC-814
Supplier:  G-Biosciences
Frequently Bought Together