Order Entry
Ireland
ContactUsLinkComponent
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SSMMS 55g/m² dark blue, inner sheet non-woven 67g/m² green, 120x120cm, DIN EN ISO 11607 1 * 60 items
Catalog # 129-1336
Stericlin® sterilisation double wrap, 2 sheets sealed at the edges, outer sheet SSMMS 55g/m² dark blue, inner sheet non-woven 67g/m² green, 120x120cm, DIN EN ISO 11607 1 * 60 items
Catalog # 129-1336
Frequently Bought Together