Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, single self-adhesive, with STEAM indicator, label size 70x 38.1mm, roll width 70mm x core 40mm, pack of 4 rolls of 2500 labels 1 * 10.000 items
Catalog # VEPA3FSKS675170
Supplier: VP Medical Packaging
Frequently Bought Together