Order Entry
Ireland
ContactUsLinkComponent
Rspo3 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of mouse Rspo3 in cell culture supernates, cell lysates, serum and plasma (heparin or EDTA). 1 * 1 KIT
  ABNOKA5159
 :  Abnova
Rspo3 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of mouse Rspo3 in cell culture supernates, cell lysates, serum and plasma (heparin or EDTA). 1 * 1 KIT
  ABNOKA5159
 :  Abnova
Frequently Bought Together