Order Entry
Ireland
ContactUsLinkComponent
AFM (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human afamin in cell culture sup ernates, serum and urine. 1 * 1 KIT
Catalog # ABNOKA5143
Supplier:  Abnova
AFM (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human afamin in cell culture sup ernates, serum and urine. 1 * 1 KIT
Catalog # ABNOKA5143
Supplier:  Abnova
Frequently Bought Together