VISCOSITY STANDARD-100-25 1 * 900 mL
Catalog # BROOAME-100CPS
Supplier: AMETEK
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
CONDUCTIVITY STANDARD 1.3µS/CM 1 * 250 mL
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Supelco® PESTANAL® Organic Reference Standard, Methomyl
Supplier: Merck
Supelco® Organic Reference Standard, 2-Methyl-2-Butanol
Supplier: Merck
VWR®, Density standards referenced at 20 °C
Supplier: VWR Chemicals
DIISOPROPYL ETHER 1 * 3 items
Supplier: Merck