CARTRIDGE SEP-PAK ACCELL PLUS QMA 1 * 50 items
Catalog # WATEWAT020545
Supplier: WATERS
Frequently Bought Together
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
SEP-PAK PLUS TC18 ENVIRONMEN TAL CARTRIDGES 1 * 50 items
Supplier: WATERS
Screw caps for certified screw top vials
Supplier: Agilent