Order Entry
Ireland
ContactUsLinkComponent
PROTEINSIMPLE ELEKTROLYTE TANK CAPS 20MM. RED US USED FOR ANLOYTE TANK AND THE GREY CAP IS USED FOR THE CATHOLYTE TANK 1 * 5 PAIR
Catalog # MSPP046-123
PROTEINSIMPLE ELEKTROLYTE TANK CAPS 20MM. RED US USED FOR ANLOYTE TANK AND THE GREY CAP IS USED FOR THE CATHOLYTE TANK 1 * 5 PAIR
Catalog # MSPP046-123
Frequently Bought Together