Human Recombinant IFNg (from E. coli)
Product Details & Documents
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Human IFN-γ is not cross-reactive with mouse IFN-γ.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
Specifications
- Pack type:Vial
- Pk:1 mg
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:16,9 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Protein function:Cytokine
- Gene ID:P01579
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Human
- Protein/peptide name:IFNg
- Protein synonyms:Type II interferon|T cell interferon|MAF|Immune Interferon
- Endotoxin level:Low
- Grade:RUO
- Sequence:MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ
- Biological activity:Bioactivity assay: Viral challenge assay using EMC virus on A549 cells, ED50 ≤0,75 ng/ml, Bioactivity ≥1,3×10⁶ units/mg
- Shipping temperature:RT
- Cat. No.:SHBT100-77-1MG
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:N
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



