Order Entry
Ireland
ContactUsLinkComponent
CMS-periphery, consisting of: mixing vessel 1 l (clamp DN ¾ ), powder funnel made of PE, fixing device for Micro-Plant, 3 x two-way ball valve, circulation line with accessories 1 * 1 items
Catalog # IKAAU083136
Supplier:  IKA
CMS-periphery, consisting of: mixing vessel 1 l (clamp DN ¾ ), powder funnel made of PE, fixing device for Micro-Plant, 3 x two-way ball valve, circulation line with accessories 1 * 1 items
Catalog # IKAAU083136
Supplier:  IKA
Frequently Bought Together