Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISOCOLOR Powder Pillows free Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 100 determinati ons 1 * 100 items
Catalog # MANA936220
Supplier:
MACHEREY-NAGEL
SDS
Expand image
VISOCOLOR Powder Pillows free Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 100 determinati ons 1 * 100 items
Catalog # MANA936220
Supplier:
MACHEREY-NAGEL
SDS
Related Information
Documentation
IFU_Viso PP Free Chlorine, 100.pdf
SDS
SDS_Viso PP Free Chlorine, 100.pdf
Documentation
Frequently Bought Together
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
VISOCOLOR Powder Pillows total Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 100 determinat ions 1 * 100 items
Supplier: MACHEREY-NAGEL