20 ml PrecisionPLUS qPCR Master Mix with ROX premixed with SYBRgreen (20x 1ml) 1 * 2.000 Tests
ZPPLUSRSY20ML
20 ml PrecisionPLUS qPCR Master Mix with ROX premixed with SYBRgreen (20x 1ml) 1 * 2.000 Tests
ZPPLUSRSY20ML
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection