Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human EpCAM TROP1 Protein, Fc Tag,Human EpCAM, Fc Tag (EPM-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG1 Fc tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBEPM-H5254-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human EpCAM TROP1 Protein, Fc Tag,Human EpCAM, Fc Tag (EPM-H5254) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a human IgG1 Fc tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBEPM-H5254-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together