Order Entry
Ireland
ContactUsLinkComponent
Nikon 100X CFI E Plan Achromat Oil Objective Lens – Numerical Aperture 1.25, Working Distance 0.23mm 1 * 1 items
Catalog # NIKOMRP71901
Supplier:  NIKON
Nikon 100X CFI E Plan Achromat Oil Objective Lens – Numerical Aperture 1.25, Working Distance 0.23mm 1 * 1 items
Catalog # NIKOMRP71901
Supplier:  NIKON
Frequently Bought Together