Order Entry
Ireland
ContactUsLinkComponent
Viscometer therm., similar Engler, similar DIN 12785, 0+55:0,2C, enclosed scale, capillary tube specially coated inside, with non- wetting blue special liquid, 300x6mm, 90mm immersion, with swelling, Richter-top finish, without button, null
Catalog # AMARM238040-CB
Supplier:  Amarell
Viscometer therm., similar Engler, similar DIN 12785, 0+55:0,2C, enclosed scale, capillary tube specially coated inside, with non- wetting blue special liquid, 300x6mm, 90mm immersion, with swelling, Richter-top finish, without button, null
Catalog # AMARM238040-CB
Supplier:  Amarell
Frequently Bought Together