Order Entry
Ireland
ContactUsLinkComponent
Extreme precision therm., with saddle, 0+100:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, 4 00x8-9mm, ganz eintauchend, Richterverschluss mit Knopf, null
Catalog # AMARL53128-CB
Supplier:  Amarell
Extreme precision therm., with saddle, 0+100:0,2C, capillary tube specially coated inside, with non-wetting blue special liquid, 4 00x8-9mm, ganz eintauchend, Richterverschluss mit Knopf, null
Catalog # AMARL53128-CB
Supplier:  Amarell
Frequently Bought Together