Order Entry
Ireland
ContactUsLinkComponent
Globe therm., complete with hollow copper ball, black, matt finish, diameter 152mm, enclosed scale therm., DIN 12785, 0+105:0,5C, c ap. lens front, colourless, red special liquid, in slotted brass case, screwed on, with ring for suspension, suitable 1 * 1 items
Catalog # AMARK194330-FL
Supplier:  Amarell
Globe therm., complete with hollow copper ball, black, matt finish, diameter 152mm, enclosed scale therm., DIN 12785, 0+105:0,5C, c ap. lens front, colourless, red special liquid, in slotted brass case, screwed on, with ring for suspension, suitable 1 * 1 items
Catalog # AMARK194330-FL
Supplier:  Amarell
Frequently Bought Together