Order Entry
Ireland
ContactUsLinkComponent
Soil therm., similar DIN 58655, -25+60:0,2C, built-in length 20mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150 bent behind, null
Catalog # AMARK188000-CB
Supplier:  Amarell
Soil therm., similar DIN 58655, -25+60:0,2C, built-in length 20mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150 bent behind, null
Catalog # AMARK188000-CB
Supplier:  Amarell
Frequently Bought Together