Order Entry
Ireland
ContactUsLinkComponent
Detect Mouse TIMP4 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. 3002190000 CN 0.1 0.045 kg Yes 365 N 20 T2 N/A 1 * 1 KIT
Catalog # BSBTEK1335
Supplier:  Boster Bio
undefined
Detect Mouse TIMP4 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. 3002190000 CN 0.1 0.045 kg Yes 365 N 20 T2 N/A 1 * 1 KIT
Catalog # BSBTEK1335
Supplier:  Boster Bio