Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

1,9-Pyrazoloanthrone ≥99%

Bottle, 300 mg
Catalog # APOSBIS1100-300MG
Irritant
Supplier:   Apollo Scientific
1,9-Pyrazoloanthrone ≥99%
Catalog # APOSBIS1100-300MG
Supplier:  Apollo Scientific
CAS Number:  129-56-6
1,9-Pyrazoloanthrone ≥99%

Bottle, 300 mg

CAS Number:  129-56-6
Price
219,00 €
Log in for availability
1 * 300 mg
Product Details & Documents

Novel, potent and selective JNK-1,-2, and -3 inhibitor (Ki = 0.19 uM). SP600125 is an ATP-competitive reversible inhibitor with >20-fold selectivity vs. a range of kinases and enzymes tested. Proc. Natl. Acad. Sci. USA 98: 13681-13686 (2001).

Synonyms:  JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
  • CAS Number:129-56-6
  • Formula:C₁₄H₈N₂O
  • MDL Number:MFCD00022289
  • Molecular Weight:220.231 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    300 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.