Order Entry
Ireland
ContactUsLinkComponent
Human B7-1, His Tag (c-terminus) is expressed from human 293 cells. It contains AA Val 35 - Asn 242 (Accession # NP_005182.1) 1 * 100 µG
Catalog # ACRBB71-H5228100UG
Supplier:  ACRO Biosystems
Human B7-1, His Tag (c-terminus) is expressed from human 293 cells. It contains AA Val 35 - Asn 242 (Accession # NP_005182.1) 1 * 100 µG
Catalog # ACRBB71-H5228100UG
Supplier:  ACRO Biosystems
Frequently Bought Together