Order Entry
Ireland
ContactUsLinkComponent
Universal test tube rack with retention system. Alphanumerically marked positions for specific identification. PP coloured red. For 40 tubes up to Ø 20 mm. Dimensions 246 x 104 x 64 mm. 1 * 1 items
Catalog # BURK9635-0032
Supplier:  BURKLE
Universal test tube rack with retention system. Alphanumerically marked positions for specific identification. PP coloured red. For 40 tubes up to Ø 20 mm. Dimensions 246 x 104 x 64 mm. 1 * 1 items
Catalog # BURK9635-0032
Supplier:  BURKLE
Frequently Bought Together