Order Entry
Ireland
ContactUsLinkComponent
TitroLine® 5000, Basic unit without magnetic stirrer, without stand rod, without mouse, without titration clamp and without electrod e, with ready to use with 50 ml dosing unit TZ 3130 with tubes, screw threads and titration tip, with external power 1 * 1 items
Catalog # SCOI285225750
Supplier:  SI Analytics
TitroLine® 5000, Basic unit without magnetic stirrer, without stand rod, without mouse, without titration clamp and without electrod e, with ready to use with 50 ml dosing unit TZ 3130 with tubes, screw threads and titration tip, with external power 1 * 1 items
Catalog # SCOI285225750
Supplier:  SI Analytics
Frequently Bought Together