Order Entry
Ireland
ContactUsLinkComponent
One block valve for connecting FlowLine electrodes to the reservoir for electrolyte solution 1 * 1 items
Catalog # SCOI285123280
Supplier:  SI Analytics
One block valve for connecting FlowLine electrodes to the reservoir for electrolyte solution 1 * 1 items
Catalog # SCOI285123280
Supplier:  SI Analytics