Order Entry
Ireland
ContactUsLinkComponent
Vials, MRQ Maximum Recovery, 1.2ml 9-425 12x32mm screw top. Caps sold separately, AQ Brand with RSA glass, reduced surface activity , SILANIZED. 100/pack. 1 * 100 items
Catalog # MICS9512S-0CV-TRSD
Supplier:  MICROSOLV
undefined
Vials, MRQ Maximum Recovery, 1.2ml 9-425 12x32mm screw top. Caps sold separately, AQ Brand with RSA glass, reduced surface activity , SILANIZED. 100/pack. 1 * 100 items
Catalog # MICS9512S-0CV-TRSD
Supplier:  MICROSOLV
Frequently Bought Together