Order Entry
Ireland
ContactUsLinkComponent
CHROMIUM 1 µG/ML 2% HCL PURE 125 ML 1 * 125 mL
Catalog # PERCN9304218
Supplier:  PerkinElmer
CHROMIUM 1 µG/ML 2% HCL PURE 125 ML 1 * 125 mL
Catalog # PERCN9304218
Supplier:  PerkinElmer
Frequently Bought Together