Order Entry
Ireland
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-12

Mouse Recombinant IL-12

100 µG
Catalog # STMC78028
Marketsource
Mouse Recombinant IL-12
Catalog # STMC78028
Marketsource
CAS Number:  
Mouse Recombinant IL-12
Mouse Recombinant IL-12

100 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 µG
Product Details & Documents

Interleukin 12 (IL-12p70) is a heterodimeric cytokine composed of p35 and p40 subunits. IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils and B cells in response to bacterial products and cytokines such as IFN-γ. The IL-12 receptor is expressed on T, NK, and dendritic cells. Upon binding, IL-12 initiates signaling via the JAK/STAT signaling pathway and stimulates NK, B, and T cells to produce IFN-γ (Watford et al.). It also regulates cytokine synthesis, proliferation of T and NK cells, and stimulates differentiation of CD4+ and CD8+ T cells (Germann and Rüde). Mice that are deficient in IL-12 are susceptible to many intracellular pathogens and have impaired IFN-γ secretion, Th1 differentiation and NK cytolytic activity; however, Th2 development and IL-4 production are enhanced (Watford et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78028
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-12
  • Purity:
    ≥95%
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.