Order Entry
Ireland
ContactUsLinkComponent
BOROSILCATE GLASS CAPILLARY FOR USE AS A SAMPLE HOLDER ON THE DIRECT SAMPLE ANALYSIS (DSA) ION SOURCE FOR AXION 2 TOF MS. FOR HOLDIN G SEMI-SOLIDS AND POWDERS. 1 * 1 items
Catalog # PERCMZ305385
Supplier:  PerkinElmer
BOROSILCATE GLASS CAPILLARY FOR USE AS A SAMPLE HOLDER ON THE DIRECT SAMPLE ANALYSIS (DSA) ION SOURCE FOR AXION 2 TOF MS. FOR HOLDIN G SEMI-SOLIDS AND POWDERS. 1 * 1 items
Catalog # PERCMZ305385
Supplier:  PerkinElmer
Frequently Bought Together