Order Entry
Ireland
ContactUsLinkComponent
THE BAFFLED CYCLONIC SPRAY CHAMBER FEATURES A CENTRAL TRANSFER TUBE WHICH ACTS AS A SECONDARY PARTICLE SEPARATOR, HELPING SEPARATE L ARGER AEROSOL PARTICLES FROM THE SAMPLE. THIS REDUCES SOLVENT LOAD IN THE PLASMA WITHOUT COMPROMISING DETECTION LIMIT 1 * 1 items
Catalog # PERCN8121060
Supplier:  PerkinElmer
THE BAFFLED CYCLONIC SPRAY CHAMBER FEATURES A CENTRAL TRANSFER TUBE WHICH ACTS AS A SECONDARY PARTICLE SEPARATOR, HELPING SEPARATE L ARGER AEROSOL PARTICLES FROM THE SAMPLE. THIS REDUCES SOLVENT LOAD IN THE PLASMA WITHOUT COMPROMISING DETECTION LIMIT 1 * 1 items
Catalog # PERCN8121060
Supplier:  PerkinElmer
Frequently Bought Together