Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Test Sieve CEREALS SS 200x32 mm/w- 2.00x20 mm 1 * 1 items
Catalog # HAVE206160270
Supplier:
NEXOPART
Expand image
Test Sieve CEREALS SS 200x32 mm/w- 2.00x20 mm 1 * 1 items
Catalog # HAVE206160270
Supplier:
NEXOPART
Frequently Bought Together
Disposable weighing dishes
Supplier: LUDI
VWR®, Aluminium foil in dispensers
Supplier: VWR Collection
VWR®, Disposable Weighing and Drying Dishes or Pans
Supplier: VWR Collection
VWR®, Magnetic Stirring Bars, Plain, Economy
Supplier: VWR Collection
VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
Spatula set
Supplier: RSG
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Supplier: Dr. Weigert
VWR®, Magnetic stirring bars, plain
Supplier: VWR Collection
VWR®, Disposable Weighing Dishes
Supplier: VWR Collection
VWR®, Magnetic stirring bars, oval
Supplier: VWR Collection
Pharmacist's spoon
Supplier: RSG