Order Entry
Ireland
ContactUsLinkComponent
Alarm function for overheating - 230 V, 400 V option model, Switchable acoustic alarm, with adjustable limit value on the independen t temperature safety device 1 * 1 items
Catalog # BIND8012-1299
Supplier:  Binder
Alarm function for overheating - 230 V, 400 V option model, Switchable acoustic alarm, with adjustable limit value on the independen t temperature safety device 1 * 1 items
Catalog # BIND8012-1299
Supplier:  Binder
Frequently Bought Together