Orange cone washers, 1.1 mm ID, fits 1.0-1.1 mm OD glass, 10 PCS, for HLU holder. 1 * 10 items
MLDV1-HLC-11
Orange cone washers, 1.1 mm ID, fits 1.0-1.1 mm OD glass, 10 PCS, for HLU holder. 1 * 10 items
MLDV1-HLC-11
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection