ASSAYCOMPLETE PUROMYCIN 1 * 1 mL
Catalog # DISX92-0028
Supplier: DISCOVERX
Frequently Bought Together
Internal thread cryogenic vials
Supplier: Corning
Bioreactors, Wheaton® CELLine™
Supplier: DWK Life Sciences
assay complete thawing reagent T3 (2 x 20 ml) 1 * 100 mL
Supplier: DISCOVERX
FalconRegistered Mark Round-Bottom Tubes, 14 mL 1 * 500 items
Supplier: STEMCELL Technologies
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Hybridisation bottles, WHEATON®
Supplier: DWK Life Sciences
Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
VWR® Cryoboxes, Double Size, 133x257 mm
Supplier: VWR Collection
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Vial centrifuge ster pp 50ml 30x115mm 1 * 25 items
Supplier: SEMADENI
PCR plates, 96-well, Armadillo™
Supplier: Thermo Fisher Scientific