Boat-combustion Quartz glass in application temperatures of up to 1050C (maximum 1600C), without handle, translucent, high chemic al resistance and purity. length 78mm, width 17mm, height 11mm. The durability or operating cycles of our Boat-combust 1 * 1 items
Catalog # TSLA73370
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together