Order Entry
Ireland
ContactUsLinkComponent
Capsule-tobacco ashin Quartz glassg in application temperatures of up to 1050C (maximum 1600C), translucent, high chemical resis tance and purity. Length 78mm, width 56mm, height 285mm. The durability or operating cycles of our Capsule Quartz glass 1 * 1 items
  TSLA73027
Capsule-tobacco ashin Quartz glassg in application temperatures of up to 1050C (maximum 1600C), translucent, high chemical resis tance and purity. Length 78mm, width 56mm, height 285mm. The durability or operating cycles of our Capsule Quartz glass 1 * 1 items
  TSLA73027
Frequently Bought Together