This product is prepared by labeling goat anti-llama IgG (H+L) with horseradish peroxidase. HRP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 1 mg
Catalog # 20475-1MG
Supplier: Biotium
Frequently Bought Together