HEPA air filter - on fresh air supply for unit filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 µm) 1 * 1 items
BIND8012-1050
: Binder
HEPA air filter - on fresh air supply for unit filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 µm) 1 * 1 items
BIND8012-1050
: Binder
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection