Order Entry
Ireland
ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed goat anti-rat IgG (H+L) with horseradish peroxidase. To minimize cross-re activity, the antibody has been adsorbed against human, bovine, horse, and rabbit serum. 1 * 1 mg
Catalog # 20406-1MG
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed goat anti-rat IgG (H+L) with horseradish peroxidase. To minimize cross-re activity, the antibody has been adsorbed against human, bovine, horse, and rabbit serum. 1 * 1 mg
Catalog # 20406-1MG
Supplier:  Biotium
Frequently Bought Together