Order Entry
Ireland
ContactUsLinkComponent
6MM PFA SECURE NUT FOR QUARTZ CYCLONIC SPRAY CHAMBERS, REVISION B - SECURES NEBULIZER TO ESI CHAMBER [ES-3199-0001-RB]. 1 * 1 items
Catalog # PERCN8141420
Supplier:  PerkinElmer
6MM PFA SECURE NUT FOR QUARTZ CYCLONIC SPRAY CHAMBERS, REVISION B - SECURES NEBULIZER TO ESI CHAMBER [ES-3199-0001-RB]. 1 * 1 items
Catalog # PERCN8141420
Supplier:  PerkinElmer
Frequently Bought Together