Order Entry
Ireland
ContactUsLinkComponent
BLACK PEEK THREADED ADAPTER TO CONVERT OPTIMA 8000 SERIES SAPPHIRE INJECTORS WITH ESI CONNECTOR FOR USE WITH OPTIMA 7X00 AND EARLIER MODELS. 1 * 1 items
Catalog # PERCN0782020
Supplier:  PerkinElmer
BLACK PEEK THREADED ADAPTER TO CONVERT OPTIMA 8000 SERIES SAPPHIRE INJECTORS WITH ESI CONNECTOR FOR USE WITH OPTIMA 7X00 AND EARLIER MODELS. 1 * 1 items
Catalog # PERCN0782020
Supplier:  PerkinElmer
Frequently Bought Together