Order Entry
Ireland
ContactUsLinkComponent
CONSUMABLES FOR APPROXIMATELY 2500 DIGESTIONS WITH HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSELS. INCLUDES 10 PRESSURE SEALS (N31 33023), 7 RUPTURE CAP DISCS (N3133022), 50 PEEK RING NUTS (N3133011) AND 5 LIP SEAL RINGS OF THE GAS CONTAINMENT MANI 1 * 1 KIT
Catalog # PERCN3133024
Supplier:  PerkinElmer
CONSUMABLES FOR APPROXIMATELY 2500 DIGESTIONS WITH HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSELS. INCLUDES 10 PRESSURE SEALS (N31 33023), 7 RUPTURE CAP DISCS (N3133022), 50 PEEK RING NUTS (N3133011) AND 5 LIP SEAL RINGS OF THE GAS CONTAINMENT MANI 1 * 1 KIT
Catalog # PERCN3133024
Supplier:  PerkinElmer
Frequently Bought Together