Order Entry
Ireland
ContactUsLinkComponent
Goat anti-Rabbit IgG (H&L), F(ab)'2 Fragment, min. cross-reactivity to non-immunoglobulin rabbit serum proteins and human serum prot eins 1 * 1 mg
Catalog # AGRIAS163530
Supplier:  Agrisera
Goat anti-Rabbit IgG (H&L), F(ab)'2 Fragment, min. cross-reactivity to non-immunoglobulin rabbit serum proteins and human serum prot eins 1 * 1 mg
Catalog # AGRIAS163530
Supplier:  Agrisera