Order Entry
Ireland
ContactUsLinkComponent
Jalas Shoes Occupational Mens 2112 Ronald, Regular Fit, Full Grain Leather Upper, PU Outsole, Oil-resistant Outsole, anti-static pro perties, double shock absorp zones, ESD, Black 1 * 1 PAIR
Catalog # EJEN2112-45
Supplier:  EJENDALS
Jalas Shoes Occupational Mens 2112 Ronald, Regular Fit, Full Grain Leather Upper, PU Outsole, Oil-resistant Outsole, anti-static pro perties, double shock absorp zones, ESD, Black 1 * 1 PAIR
Catalog # EJEN2112-45
Supplier:  EJENDALS